Learn More
Aviva Systems Biology Corp TCEAL1 Antibody - middle region (ARP35748_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP35748_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TCEAL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TCEAL1. Alternate names: SIIR, p21, pp21ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55837