Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCEAL1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $352.89

Specifications

Antigen TCEAL1
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB125649
View Documents
Novus Biologicals
NBP310374100UL
100 μg
Each for $352.89
Add to cart
 
NB125650
View Documents
Novus Biologicals
NBP31037425UL
25 μg
Each for $106.19
Add to cart
 
Description

Description

TCEAL1 Polyclonal antibody specifically detects TCEAL1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

TCEAL1
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Purified
RUO
PBS buffer, 2% sucrose
9338
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin
Unconjugated
Rabbit
Human
Nuclear phosphoprotein p21/SIIR, p21, pp21, SIIRTCEA-like protein 1, transcription elongation factor A (SII)-like 1, transcription elongation factor A protein-like 1, Transcription elongation factor S-II protein-like 1
The immunogen is a synthetic peptide directed towards the middle region of human TCEAL1 (NP_001006641). Peptide sequence EGLSRKDLFEGRPPMEQPPCGVGKHKLEEGSFKERLARSRPQFRGDIHGR
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit