Learn More
Aviva Systems Biology Corp TCEAL3 Antibody - N-terminal region (ARP50724_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP50724_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TCEAL3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TCEAL3. Alternate names: MGC15737ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60418