Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCEP-HCl, Molecular biology grade, OmniPur™, Calbiochem™,
Click to view available options
Quantity:
2g
Packaging:
Glass Ampoule
Specifications
Packaging | Glass Ampoule |
Color | White |
Quantity | 2g |
CAS | 51805-45-9 |
Molecular Formula | C9H12O6P |
Synonym | tris 2-carboxyethyl phosphine hydrochloride,3,3',3-phosphinetriyltripropanoic acid hydrochloride,tcep hcl,tcep hydrochloride,tcep,unii-h49aam893k,tris carboxyethyl phosphine hydrochloride,tris-2-carboxyethyl phosphine hydrochloride,tris 2-carboxyethyl phosphine hcl,3-bis 2-carboxyethyl phosphanyl propanoic acid hydrochloride |
InChI Key | PZBFGYYEXUXCOF-UHFFFAOYSA-K |
Molecular Weight (g/mol) | 247.16 |
PubChem CID | 2734570 |
Physical Form | Crystalline powder |
Show More |