Learn More
Aviva Systems Biology Corp TCF4 Antibody - N-terminal region (P100775_P050) | 100uL
Supplier:  Aviva Systems Biology Corp P100775_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TCF4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TCF4. Alternate names: E2-2, ITF2, SEF2, SEF2-1, SEF2-1A, SEF2-1B, PTHS, ITF-2, SEF-2, TCF-4, bHLHb19ApplicationWB. Species reactivity: Guinea Pig, Horse, Human, Mouse
Catalog No. 50-014-62708