Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCF4 Mouse anti-Human, Clone: 6H5.3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP266586

Catalog No. NBP266586


Add to cart

Description

Description

TCF4 Monoclonal antibody specifically detects TCF4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
Specifications

Specifications

TCF4
6H5.3
Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500
Purified
TCF4
Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with recombinant protein corresponding to amino acids 31-331 of the human TCF-4 protein.
Protein A or G purified
Transcription Factors and Regulators
Primary
6925
Immunohistochemistry, Immunoprecipitation, Western Blot
Unconjugated
0.7M tris-glycine, pH 7.4, 0.105M NaCl, 30% glycerol with 0.035% Sodium Azide
BHLHB19, bHLHb19E2-2, Class B basic helix-loop-helix protein 19, Immunoglobulin transcription factor 2, ITF-2, ITF2PTHS, MGC149724, SEF-2, SEF2-1, SEF2-1A, SEF2-1B, SEF2MGC149723, SL3-3 enhancer factor 2, TCF-4, transcription factor 4
Mouse
IgG2a
100μg
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
Human, Monkey, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit