Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCF7L1/TCF3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP310886100UL

 View more versions of this product

Catalog No. NB126673


Add to cart

Description

Description

TCF7L1/TCF3 Polyclonal specifically detects TCF7L1/TCF3 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

TCF7L1/TCF3
Unconjugated
PBS buffer, 2% sucrose
HMG box transcription factor 3, TCF3, TCF-3, transcription factor 7-like 1, transcription factor 7-like 1 (T-cell specific, HMG-box)
The immunogen is a synthetic peptide directed towards the N terminal region of human TCF7L1/TCF3 (NP_112573). Peptide sequence NDELIPFQDEGGEEQEPSSDSASAQRDLDEVKSSLVNESENQSSSSDSEA
100 μg
Core ESC Like Genes, Signal Transduction, Stem Cell Markers, Wnt Signaling Pathway
Primary
83439
Western Blot, Immunohistochemistry
Western Blot 1.0 ug/ml, Immunohistochemistry
Purified
Rabbit
Affinity purified
RUO
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit