Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCFL5 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA124394
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA124394 50 μg
1 options

Catalog No. PIPA124394

Supplier: Thermo Scientific PA124394

Rabbit Polyclonal Antibody

PA1-24394 detects TCFL5 in Human samples. PA1-24394 has been successfully used in Western Blot procedures. PA1-24394 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human TCFL5. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

TCFL5 is a new bHLH transcription factor that negatively regulates upstream transcription factor-dependent transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCFL5
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Gene TCFL5
Gene Accession No. Q9UL49
Gene Alias CHA, E2BP-1, Figlb, MGC46135
Gene Symbols TCFL5
Host Species Rabbit
Immunogen Synthetic peptide region of human TCFL5.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10732
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.