Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TCHHL1 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518970
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.
This gene belongs to the S100 fused-type protein (SFTP) gene family, and is located in a cluster of SFTP genes on chromosome 1q21. Several members of this family have been implicated in the development of complex skin disorders. This gene is evolutionarily conserved; its expression appears to be hair-specific and spatially restricted within the distal inner root sheath of the hair follicle. It thus may have an important role in hair morphogenesis. [provided by RefSeq, Aug 2013].
Specifications
TCHHL1 | |
Polyclonal | |
Unconjugated | |
TCHHL1 | |
basalin; intermediate filament-associated protein; Protein S100-A17; S100 calcium binding protein A17; S100 calcium-binding protein A17; S100A17; TCHHL1; THHL1; trichohyalin like 1; trichohyalin-like 1; trichohyalin-like protein 1 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
126637 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q5QJ38 | |
TCHHL1 | |
Synthetic peptide sequence (QALEDKQGHPQRE) corresponding to the internal amino acids of TCHHL1 (aa 879-892). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction