Learn More
TCI OPTI-GUARD Fit CN, 3/pk , TCI America™
Manufacturer:  TCI America S14443CLM
Catalog No. S14443CLM
Add to cart
Description
Description
Universal filling for most columns and zero dead-volume connection without any effect on column performance. This series is recommended for the further purification of relatively clean samples using either 4.6 mm I.D. or 2.0 mm I.D. HPLC columns.
Specifications
Specifications
1 mm
Cyano
15 mm