Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCL1 Mouse anti-Human, Clone: 1 to 21, BD Pharmingen™

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 565855

Catalog No. BDB565855


Add to cart

Description

Description

TCL1 Monoclonal antibody specifically detects TCL1 in Human samples. It is validated for Flow cytometry (Intracellular Staining)) .
Specifications

Specifications

TCL1
1 to 21
Unconjugated
Purified
Murine
Mouse BALB/c IgG2b, κ
RUO
Store undiluted at 4°C.
Monoclonal
Flow Cytometry, Flow Cytometry (Intracellular Staining)
0.5mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Oncogene TCL-1; Tcl1; Tcl1a; T-cell leukemia/lymphoma protein 1A
Human TCL1 Peptide
0.1mg
Cell Biology
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Reseach Use Only.