Learn More
Aviva Systems Biology Corp TCL1A Antibody - N-terminal region (ARP58808_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP58808_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TCL1A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TCL1A. Alternate names: TCL1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-62430