Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCL1A Mouse anti-Human, DyLight™ 650, Clone: 2019-01-21 00:00:00, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP143253C

Catalog No. NBP143253C


Add to cart

Description

Description

TCL1A Monoclonal antibody specifically detects TCL1A in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

TCL1A
2019-01-21 00:00:00
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
TCL1A
The immunogen for this antibody was TCL1.
Protein A or G purified
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
DyLight™ 650
50 mM sodium borate with 0.05% sodium azide
Oncogene TCL1, Oncogene TCL-1, Protein p14 TCL1, T-cell leukemia/lymphoma 1A, T-cell lymphoma-1, T-cell lymphoma-1A, TCL1T-cell leukemia/lymphoma protein 1A
Mouse
IgG2b Kappa
0.1 mL
Primary
8115.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit