Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCN 213, Tocris Bioscience™

Catalog Number p-4816921
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
556803-08-8
C18H24N4OS2
376.537
XBAZPYFIYYCZBO-UHFFFAOYSA-N
2-5-benzylamino-1,3,4-thiadiazol-2-yl sulfanyl-n-cyclohexylmethyl acetamide, 2-5-benzylamino-1,3,4-thiadiazol-2-yl thio-n-cyclohexylmethyl acetamide, 2-5-benzylamino-1,3,4-thiadiazol-2-ylthio-n-cyclohexylmethyl acetamide, n-cyclohexylmethyl-2-5-phenylmethyl amino-1,3,4-thiadiazol-2-yl thio acetamide
2608149
2-[[5-(benzylamino)-1,3,4-thiadiazol-2-yl]sulfanyl]-N-(cyclohexylmethyl)acetamide
C1CCC(CC1)CNC(=O)CSC2=NN=C(S2)NCC3=CC=CC=C3
This item is not returnable. View return policy
556803-08-8
C18H24N4OS2
376.537
XBAZPYFIYYCZBO-UHFFFAOYSA-N
2-5-benzylamino-1,3,4-thiadiazol-2-yl sulfanyl-n-cyclohexylmethyl acetamide, 2-5-benzylamino-1,3,4-thiadiazol-2-yl thio-n-cyclohexylmethyl acetamide, 2-5-benzylamino-1,3,4-thiadiazol-2-ylthio-n-cyclohexylmethyl acetamide, n-cyclohexylmethyl-2-5-phenylmethyl amino-1,3,4-thiadiazol-2-yl thio acetamide
2608149
2-[[5-(benzylamino)-1,3,4-thiadiazol-2-yl]sulfanyl]-N-(cyclohexylmethyl)acetamide
C1CCC(CC1)CNC(=O)CSC2=NN=C(S2)NCC3=CC=CC=C3

NMDA antagonist; selective for GluN2A over GluN2B containing receptors

NMDA receptor antagonist, selective for GluN1/GluN2A (formally NR1/NR2A) (pIC50 = 5.4) over GluN1/GluN2B (formally NR1/NR2B) containing receptors.

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

Quantity 10 mg
Formula Weight 376.54
Percent Purity >99%
Chemical Name or Material TCN 213
Recommended Storage Store at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit