TCN 213, Tocris™

NMDA antagonist; selective for NR2A over NR2B

Manufacturer: R&D Systems 4163/10

Quantity 10mg

 View more versions of this product

Catalog No. 41-631-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >99%
    Molecular Formula C18H24N4OS2
    Formula Weight 376.54
    Synonym N-(Cyclohexylmethyl)-2-[(5-[(phenylmethyl)amino]-1, 3, 4-thiadiazol-2-yl}thio]acetamide
    Chemical Name or Material TCN 213

    Specifications

    Formula Weight 376.54
    Quantity 10mg
    Assay Percent Range >99%
    Molecular Formula C18H24N4OS2
    CAS 556803-08-8
    Synonym N-(Cyclohexylmethyl)-2-[(5-[(phenylmethyl)amino]-1, 3, 4-thiadiazol-2-yl}thio]acetamide
    Recommended Storage Store at +4°C
    Chemical Name or Material TCN 213

    Store at +4°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit