Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TCN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576937
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Transcobalamin I (TCI) and Transcobalamin II (TCII) are secreted proteins belonging to the eukaryotic cobalamin transport proteins family and also to the vitamin B12-binding protein family. The genes encoding these proteins map to chromosome 11q11-q12 and 22q12.2, respectively. Transcobalamin I is a constituent of secondary granules in neutrophils, while Transcobalamin II binds cobalamin and mediates its transport into cells. These plasma proteins are expressed in various tissues and secretions.
Specifications
TCN1 | |
Polyclonal | |
Unconjugated | |
TCN1 | |
haptocorin; haptocorrin; HC; protein R; TC I; TC1; TC-1; TCI; TCN1; transcobalamin 1; Transcobalamin I; transcobalamin I (vitamin B12 binding protein, R binder family); transcobalamin-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6947 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P20061 | |
TCN1 | |
Recombinant full length Human TCN1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction