Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TCN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578194
Description
Positive Control: human plasma Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Transcobalamin I (TCI) and Transcobalamin II (TCII) are secreted proteins belonging to the eukaryotic cobalamin transport proteins family and also to the vitamin B12-binding protein family. The genes encoding these proteins map to chromosome 11q11-q12 and 22q12.2, respectively. Transcobalamin I is a constituent of secondary granules in neutrophils, while Transcobalamin II binds cobalamin and mediates its transport into cells. These plasma proteins are expressed in various tissues and secretions.
Specifications
TCN1 | |
Polyclonal | |
Unconjugated | |
TCN1 | |
haptocorin; haptocorrin; HC; protein R; TC I; TC1; TC-1; TCI; TCN1; transcobalamin 1; Transcobalamin I; transcobalamin I (vitamin B12 binding protein, R binder family); transcobalamin-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6947 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P20061 | |
TCN1 | |
Recombinant protein encompassing a sequence within the center region of human TCN1. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction