Learn More
TCP-1 beta Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA593047
Description
Immunogen sequence: GGSLADSYLD EGFLLDKKIG VNQPKRIENA KILIANTGMD TDKIKIFGSR VRVDSTAKVA EIEHAEKEKM KEKVERILKH GINCFINRQL IYNYPEQLFG AAGVMAIEHA DFAGVERLAL VTGGEIASTF DHPELVKLGS CKLIEEVMIG EDKLIHFSGV ALGEACTIVL RGATQQILDE AERSLHDALC VLAQTVKDSR TVYGGGCSEM LMAHAVTQLA NRTPGKEAVA MESYAKALRM LPTIIADNAG YDSADLVAQL RAAHSEGNTT AGLDMREGTI GDMAILGITE SFQVKRQVLL SAAEAAEVIL RVDNIIKAAP RKRVPDHHPC; Positive Samples: MCF7, SKOV3, A-549, HT-29, Mouse spleen, Mouse ovary, Mouse heart; Cellular Location: Cytoplasm
TCP-1 beta is a molecular chaperone that is member of the chaperonin containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). The CCT complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternate transcriptional splice variants of the TCP-1 beta gene have been observed but not thoroughly characterized. A disease associated with TCP-1 beta protein dysfunction include epididymo-orchitis.Specifications
TCP-1 beta | |
Polyclonal | |
Unconjugated | |
CCT2 | |
CCT2 | |
Recombit fusion protein containing a sequence corresponding to amino acids 206-535 of human CCT2 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunofluorescence, Immunoprecipitation, Western Blot | |
5.63 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P78371, P80314 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10576, 12461 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.