Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCP1 alpha Rat anti-Human, Mouse, Rat, Porcine, Bovine, Canine, C. elegans, Drosophila, Guinea Pig, Hamster, Monkey, Rabbit, Yeast, DyLight 550, Clone: 91a, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259347R

Catalog No. NBP259347R


Add to cart

Description

Description

TCP1 alpha Monoclonal antibody specifically detects TCP1 alpha in Human, Mouse, Rat, Porcine, Bovine, Canine, C. elegans, Drosophila, Guinea Pig, Hamster, Monkey, Rabbit, Yeast samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

TCP1 alpha
91a
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Purified
TCP1
Recombinant Mouse TCP1 alpha protein fragment (carboxy terminal region).
Protein G purified
Cancer, Hypoxia, Membrane Trafficking and Chaperones, Stem Cell Markers
Primary
6950
Human, Monkey, Rabbit, Yeast, Mouse, Rat, Porcine, Bovine, Canine, C. elegans, Drosophila, Guinea Pig, Hamster
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
CCT1T-complex protein 1 subunit alpha, CCTa, CCT-alpha, D6S230E, tailless complex polypeptide 1, t-complex 1, T-complex protein 1, alpha subunit, TCP-1-alpha
Rat
IgG2a
100 ul
Store at 4C in the dark.
Monoclonal
Detects 60kDa. Also detects 92kDa. Cross reactivity with human HSP60 has been observed with this in immunoblot analysis. Reacts weakly with Saccharomyces cerevisiae, consistent with the epitope sequence being AKLRS (instead of AKLRA). In C. elegans, it reacts with TCP1 alpha and another CCT subunit. In plants, it recognizes TCP1 of Pisum sativum, and the sequence of Arabidopsis thalania TCP1 over the region of the epitope AKLRA. It has also been shown that it reacts with a subunit of a specialized chaperonin which folds phytochrome.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit