Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCP1-beta Mouse anti-Drosophila, Clone: F39 P7 F11, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP266591
Description
TCP1-beta Monoclonal antibody specifically detects TCP1-beta in Drosophila samples. It is validated for Western Blot, Immunohistochemistry.Specifications
TCP1-beta | |
F39 P7 F11 | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500 | |
PBS with 0.08% Sodium Azide | |
10576 | |
Mouse | |
IgG2a | |
Primary | |
200μg | |
Store at -20°C. Avoid freeze-thaw cycles. |
Immunohistochemistry,Western Blot | |
Unconjugated | |
Purified | |
Cctb, CCT-beta, CCTBPRO1633,99D8.1, chaperonin containing t-complex polypeptide 1, beta subunit, chaperonin containing t-complex polypeptide 1, subunit 2, chaperonin containing TCP1, subunit 2 (beta), MGC142074, MGC142076, T-complex protein 1 subunit beta, T-complex protein 1, beta subunit, TCP-1-beta | |
CCT2 | |
Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with a synthetic peptide derived from the C-terminal of the TCP-1 protein and mouse myeloma Ag8563 cells. Sequence common in human, frog, mouse, rat | |
Monoclonal | |
Protein A or G purified | |
Cell Cycle and Replication, Membrane Trafficking and Chaperones | |
Drosophila,Human,Mouse,Rat,Xenopus |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title