Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tcp1, Rat anti-Mouse, PE/ATTO594, Clone: 23c, Abnova
Rat Monoclonal Antibody
Manufacturer: Abnova Corporation MAB18724
Specifications
Tcp1 | |
23c | |
Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunoprecipitation Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
t-complex protein 1 | |
AI528772|CCT|Cct1|Ccta|TRic|Tcp-1|Tp63|c-cpn|p63 | |
Rat | |
IgG2c | |
RUO | |
Antibody | |
Monoclonal | |
It detects ~60kDa. Also detects ~92kDa. The addition of an alanine (LDDA COOH) prevents binding in ELISA assays to immobilized synthetic peptide sequences. |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
PE/ATTO594 | |
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide). | |
Liquid | |
Tcp1 | |
Purified recombinant mouse TCP1 alpha construct encoding the C-terminal half of the 1.8 kb full-length Tcp 1b gene expressed in E. coli. Detects the COOH group. | |
100 μg | |
Store at -20°C. | |
Primary | |
21454 | |
Bovine, Dog, Hamster, Mouse, Rabbit, Rat, Sheep |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title