Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
TCP11 Mouse anti-Human, Clone: 2E3, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA524421
Catalog No. PIMA524421
Description
Description
Peptide Sequence: NTASLMGQLQ NIAKKENCVC SVIDQRIHLF LKCCLVLGVQ RSLLDLPGGL TLIEAELAEL GQKFVNLTHH NQQVFGPYYT EILKTLISPA QALETKVESV
May play an important role in sperm function and fertility.
Specifications
Specifications
TCP11
Monoclonal
Unconjugated
TCP11
Liquid
TCP11
TCP11 (NP_061149.1, 342 a.a. ∽ 441 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Protein A
RUO
Antibody
Monoclonal
Human
ELISA, Western Blot
2E3
PBS with no preservative; pH 7.4
Q8WWU5
D6S230E, T-complex protein 11 homolog, t-complex 11 (a murine tcp homolog)
Mouse
IgG2a, kappa
100 μg
-20° C
Primary
6954