Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR beta Armenian Hamster anti-Mouse, Pacific Blue™, Clone: H57-597, Invitrogen™ Non-distribution product as customer accommodation.

Armenian Hamster Monoclonal Antibody

Manufacturer:  Invitrogen HM3628

Catalog No. 50-113-8082


Add to cart

Description

Description

The T cell antigen receptor (TCR) consists of a ligand-specific alpha/beta heterodimer non-covalently associated with five invariant chains including the CD3 gamma/delta/eta and zeta subunits, all of which are required for efficient surface expression. T cell activation through the TCR induces cellular differentiation and/or proliferation and the production of lymphokines and cytokines. Both the CD3 and TCR zeta subunits are proposed to be responsible for the intracellular signal transduction events.
Specifications

Specifications

TCR beta, TCR alpha beta
Monoclonal
0.2 mg/mL
PBS with 0.1% sodium azide
P01849
Tcra, Tcrb, FLJ22602, MGC117436, MGC22624, MGC23964, MGC71411, TCRA, TCRD, TRA, T-cell antigen receptor, alpha polypeptide, T-cell receptor, TCR beta
Armenian Hamster
IgG
1 mL
4° C
Primary
21577
Flow Cytometry
H57-597
Pacific Blue™
Tcrb
Liquid
Tcrb
TCR affinity purified from mouse T cell hybridoma DO-11.101
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit