Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
TCR alpha Mouse anti-Human, Alexa Fluor™ 532, Clone: 34, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP252715AF532
Catalog No. NP2271A32
Description
Description
TCR alpha Monoclonal antibody specifically detects TCR alpha in Human samples. It is validated for Western Blot,Flow Cytometry,ELISA.
Specifications
Specifications
TCR alpha
34
Western Blot, Flow Cytometry, ELISA
C), D, FLJ22602, J, MGC117436, MGC22624, MGC23964, MGC71411, T cell receptor alpha locus, T-cell antigen receptor, alpha polypeptide, T-cell receptor, alpha (V, TCRA, TCRD, TRA
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
This antibody binds to the T-cell receptor alpha chains.
ELISA, Flow Cytometry, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
TRA
Purified recombinant soluble TCR from the T-cell hybridoma 85.33, corresponding to V alpha 11
0.1 mL
Primary
6955
Human