Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR alpha Rabbit anti-Human, Alexa Fluor™ 594, Clone: 34, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP252684AF594

Catalog No. NP22684A94


Add to cart

Description

Description

TCR alpha Monoclonal antibody specifically detects TCR alpha in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications

Specifications

TCR alpha
34
Western Blot, Flow Cytometry, ELISA
Conjugated, Purified
TRA
Purified recombinant soluble TCR from the T-cell hybridoma 85.33, corresponding to V alpha 11
Protein A or G purified
Adaptive Immunity, Cell Biology, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary
6955.0
Human
ELISA, Flow Cytometry, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
C), D, FLJ22602, J, MGC117436, MGC22624, MGC23964, MGC71411, T cell receptor alpha locus, T-cell antigen receptor, alpha polypeptide, T-cell receptor, alpha (V, TCRA, TCRD, TRA
Rabbit
IgG Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
This antibody binds to the T-cell receptor alpha chains.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit