Learn More
TCR β Chain Hamster anti-Mouse, APC, Clone: H57-597, BD
Hamster Monoclonal Antibody
$116.00 - $247.00
Specifications
Clone | H57-597 |
---|---|
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Host Species | Hamster |
Catalog Number | Mfr. No. | Quantity | Conjugate | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Conjugate | Price | Quantity & Availability | ||||
BDB561080
|
BD Biosciences
561080 |
25 μg | APC |
Each for $116.00
|
|
||||
BDB553174
|
BD Biosciences
553174 |
0.1 mg | APC |
Each for $247.00
|
|
||||
BDB560656
|
BD Biosciences
560656 |
50 μg | APC-Cy7 |
Each for $188.00
|
|
||||
Description
The H57-597 antibody reacts with a common epitope of the β chain of the T-cell Receptor (TCR) complex on αβ TCR-expressing thymocytes and peripheral T lymphocytes and NK1.1+ thymocytes and NK-T cells of all mouse strains tested. It does not react with γδ TCR-bearing T cells. In the fetal and adult thymus, the TCR β-chain may form homodimers or pair with the pre-TCR α-chain on the surface of immature thymocytes before expression of the TCR α-chain. Plate-bound or soluble H57-597 antibody activates αβ TCR-bearing T cells, and plate-bound mAb can induce apoptotic death.
Specifications
H57-597 | |
Flow Cytometry | |
Hamster | |
Mouse | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
0.2mg/mL | |
Monoclonal | |
RUO | |
TCR affinity-purified from mouse T-cell hybridoma DO-11.10 | |
Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.