Learn More
TCR β Chain Hamster anti-Mouse, Brilliant Violet 786, Clone: H57-597, BD Optibuild™
Armenian Hamster Monoclonal Antibody
Supplier: BD Biosciences 742484/DUP
Description
The H57-597 antibody reacts with a common epitope of theβchain of the T-cell Receptor (TCR) complex onαβTCR-expressing thymocytes, peripheral T lymphocytes, NK1.1+ thymocytes, and NK-T cells of all mouse strains tested. It does not react withγδTCR-bearing T cells. In the fetal and adult thymus, the TCRβ-chain may form homodimers or pair with the pre-TCRα-chain on the surface of immature thymocytes before TCRα-chain expression. Plate-bound or soluble H57-597 antibody activatesαβTCR-bearing T cells, and plate-bound mAb can induce apoptotic death.
The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.Specifications
TCR β Chain | |
H57-597 | |
Brilliant Violet 786 | |
Tcrb, TCRbeta, TCRβ, T cell receptor beta chain | |
TCR affinity-purified from mouse T-cell hybridoma DO-11.10 | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Armenian Hamster | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2 λ1 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.