Learn More
TCR DO11.10 Mouse anti-Mouse, FITC, Clone: KJ1-26, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MM7501
Catalog No. 50-113-8135
Add to cart
Description
Description
TCR DO11 (DO11.10) is a receptor found on lymphocytes of DO11.10 transgenic mice and T-cell receptors of BALB/c-derived DO11.10 and DO11.10.24 T cell hybridomas. The DO11.10 TCR recognizes a C-terminal ovalbumin peptide (OVA 323-339) in the context of I-Ad major histocompatibility molecules. The use of antibodies against DO11.10 have proven to be instrumental in describing many of the intricate details of T cell immunity, including T cell activation, development, and function.
Specifications
Specifications
TCR DO11.10
Monoclonal
FITC
Liquid
Mouse
IgG2a
1 mL
4° C
Primary
Mouse
Flow Cytometry
KJ1-26
PBS with 0.1% sodium azide
D011.10; T-cell receptor DO11.10
Mouse TCR Clonotype.
Purified
RUO
Antibody
Monoclonal