Learn More
Genetex IncSupplier Diversity Partner Anti-TCR gamma + delta antibody [5A6.E9] DFS Item
Manufacturer:  Genetex IncSupplier Diversity Partner GTX15594
The Anti-TCR gamma + delta antibody [5A6.E9] from GeneTex is a Mouse Monoclonal antibody to TCR gamma + delta. The Anti-TCR gamma + delta antibody [5A6.E9] Recognizes Human. The Anti-TCR gamma + delta antibody [5A6.E9] has been validated for the following Applications: Blocking, ELISA, FACS, IHC, IHC-Fr, IP, Activation.
Catalog No. NC1471669