Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR gamma/delta Mouse anti-Human, DyLight™ 650, Clone: B1.1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP143121C

Catalog No. NBP143121C


Add to cart

Description

Description

TCR gamma/delta Monoclonal antibody specifically detects TCR gamma/delta in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

TCR gamma/delta
B1.1
Flow Cytometry
Conjugated, Purified
TRG
The immunogen for this antibody was gamma delta TCR.
Protein A or G purified
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry
DyLight™ 650
50 mM sodium borate with 0.05% sodium azide
T cell receptor gamma locus, T-cell antigen receptor, gamma polypeptide, T-cell rearranging gene, gamma, T-cell receptor, gamma cluster, TCRG, TRG
Mouse
IgG1 Kappa
0.1 mL
Primary
6965.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit