Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCR gamma/delta Mouse anti-Human, PE-Cyanine7, Clone: B1.1, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP143121PECY7
Description
TCR gamma/delta Monoclonal antibody specifically detects TCR gamma/delta in Human samples. It is validated for Flow Cytometry.Specifications
TCR gamma/delta | |
B1.1 | |
Flow Cytometry | |
Conjugated, Purified | |
TRG | |
The immunogen for this antibody was gamma delta TCR. | |
Protein A or G purified | |
Store at 4°C in the dark. Do not freeze. | |
Monoclonal | |
Human |
Flow Cytometry | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
T cell receptor gamma locus, T-cell antigen receptor, gamma polypeptide, T-cell rearranging gene, gamma, T-cell receptor, gamma cluster, TCRG, TRG | |
Mouse | |
IgG1 Kappa | |
0.1 mL | |
Primary | |
6965.0 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title