Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR Vβ3 Mouse anti-Human, Brilliant Violet 480, Clone: JOVI.3, BD OptiBuild™

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 747991

Catalog No. BDB747991


Add to cart

Description

Description

TCR Vβ3 Monoclonal antibody specifically detects TCR Vβ3 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

TCR Vβ3
JOVI.3 (also known as JOVI-3)
Brilliant Violet 480
Mouse
50 μg
Refrigerate (2 to 8°C)
Monoclonal
Human
Flow Cytometry
0.2 mg/mL
Conjugated, Purified
IgG2a, κ
RUO
Primary
TCR Vβ3
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit