Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCR V alpha 2 Monoclonal Antibody (B20.1), NovaFluor™ Yellow 610, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Specifications

Antigen TCR V alpha 2
Clone B20.1
Concentration 0.1 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
M033T02Y03
Encompass
View Documents
LIFE TECHNOLOGIES
M033T02Y03
25 μg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
M033T03Y03
Encompass
View Documents
LIFE TECHNOLOGIES
M033T03Y03
100 μg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

TCR V alpha 2 Monoclonal antibody specifically detects TCR V alpha 2 in Mouse samples. It is validated for Flow Cytometry
Specifications

Specifications

TCR V alpha 2
0.1 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
IgG2a λ
Affinity chromatography
Antibody
B20.1
Flow Cytometry
NovaFluor Yellow 610
Rat
Mouse
TCRVA; TCRVA2; V alpha 2; V alpha-2 TCR; Va 2; Va2; Valpha 2; Valpha2
Primary
4° C, store in dark, DO NOT FREEZE!
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit