Learn More
TCR VBta3 Mouse, anti-Human, R718, Clone: JOVI.3, BD OptiBuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 751957
Catalog No. BDB751957
Add to cart
Description
Description
The JOVI.3 monoclonal antibody specifically recognizes the human variable beta 3 region of the T cell receptor for antigen (TCR Vβ3). The JOVI.3 antibody reportedly stains approximately 1-10% of human CD3-positive peripheral blood lymphocytes including CD4+ or CD8+ T cells. The variation in human TCR Vβ3 expression correlates with allelic polymorphism in the spacer region of recombination signal sequence. The JOVI.3 antibody is mitogenic for peripheral blood T cells expressing TCR Vβ3.
Specifications
Specifications
TCR VBta3
JOVI.3
Liquid
50 μg
Refrigerate Upon Arrival
Monoclonal
Flow Cytometry
R718
Mouse
RUO
Primary
Human