Learn More
TCRalpha Mouse anti-Human, Janelia Fluor 646, Clone: 34, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP252715JF646
Catalog No. NBP52715646
Add to cart
Description
Description
TCRalpha Monoclonal antibody specifically detects TCRalpha in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
TCRalpha
34
Flow Cytometry
Purified
TRA
Purified recombinant soluble TCR from the T-cell hybridoma 85.33, corresponding to V alpha 11
Protein A or G purified
Adaptive Immunity, Cell Biology, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary
6955
Human
Flow Cytometry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
C), D, FLJ22602, J, MGC117436, MGC22624, MGC23964, MGC71411, T cell receptor alpha locus, T-cell antigen receptor, alpha polypeptide, T-cell receptor, alpha (V, TCRA, TCRD, TRA
Mouse
IgG1 Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
This antibody binds to the T-cell receptor alpha chains.