Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCR alpha Antibody (34), Janelia Fluor™ 549 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP252684JF549
Description
TCR alpha Monoclonal specifically detects TCR alpha in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
TCRalpha | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TRA | |
Purified recombinant soluble TCR from the T-cell hybridoma 85.33, corresponding to V alpha 11 | |
0.1 mL | |
Primary | |
This antibody binds to the T-cell receptor alpha chains. | |
Store at 4°C in the dark. | |
IgG κ |
Flow Cytometry | |
34 | |
Flow Cytometry | |
C), D, FLJ22602, J, MGC117436, MGC22624, MGC23964, MGC71411, T cell receptor alpha locus, T-cell antigen receptor, alpha polypeptide, T-cell receptor, alpha (V, TCRA, TCRD, TRA | |
Rabbit | |
Protein A or G purified | |
Adaptive Immunity, Cell Biology, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
6955 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction