Learn More
TCRgamma/delta Mouse anti-Human, Janelia Fluor 646, Clone: B1.1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP143121JF646
Catalog No. NBP43121646
Add to cart
Description
Description
TCRgamma/delta Monoclonal antibody specifically detects TCRgamma/delta in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
TCRgamma/delta
B1.1
Flow Cytometry
Purified
TRG
The immunogen for this antibody was gamma delta TCR.
Protein A or G purified
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
T cell receptor gamma locus, T-cell antigen receptor, gamma polypeptide, T-cell rearranging gene, gamma, T-cell receptor, gamma cluster, TCRG, TRG
Mouse
IgG1 Kappa
0.1mL
Primary
6965