Learn More
TCS 1105, Tocris™
GABAA α2 benzodiazepine receptor agonist
Manufacturer:  R&D Systems 3942/10
Quantity 10mg
 View more versions of this product
Catalog No. 39-4210
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>99%
296.3
TCS 1105
C17H13FN2O2
N-[(-4-Fluorophenyl)methyl]-α-oxo-1H-indole-3-acetamide
Specifications
10mg
>99%
296.3
TCS 1105
185391-33-7
C17H13FN2O2
N-[(-4-Fluorophenyl)methyl]-α-oxo-1H-indole-3-acetamide
Safety and Handling
Recommended Storage : Store at Room Temperature