Learn More
Tocris Bioscience™ TCS 183
Fragment 1-13 of GSK-3β sequence
Manufacturer:  Tocris Bioscience™ 2236/1
Catalog No. 22-361-R
Add to cart
This item is not returnable. View return policy
Description
Description
Fragment 1-13 of human and mouse GSK-3β sequence. Potentially useful as a competitive inhibitor of GSK-3β (Ser9) phosphorylation.
Control Peptide also available.
Specifications
Specifications
TCS 183
1mg
TCS 183
1425.58
Met-Ser-Gly-Arg-Pro-Arg-Thr-Thr-Ser-Phe-Ala-Glu-Ser-NH2