Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCS 2002, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
1005201-24-0
C18H14N2O3S
338.381
RCKYSTKYIVULEK-UHFFFAOYSA-N
2-3-4-methanesulfinylphenyl-1-benzofuran-5-yl-5-methyl-1,3,4-oxadiazole,mmbo,1,3,4-oxadiazole, 2-methyl-5-3-4-methylsulfinyl phenyl-5-benzofuranyl,1,3,4-oxadiazole,2-methyl-5-3-4-methylsulfinyl phenyl-5-benzo b furanyl,2-3-4-methanesulfinyl phenyl-1-benzofuran-5-yl-5-methyl-1,3,4-oxadiazole,2-methyl-5-3-4-methylsulfinyl phenyl-1-benzofuran-5-yl-1,3,4-oxadiazole,2-methyl-5-3-4-methylsulfinyl phenyl-5-benzofuranyl-1,3,4-oxadiazole
24855958
2-methyl-5-[3-(4-methylsulfinylphenyl)-1-benzofuran-5-yl]-1,3,4-oxadiazole
CC1=NN=C(O1)C2=CC3=C(C=C2)OC=C3C4=CC=C(C=C4)S(=O)C
This item is not returnable. View return policy
1005201-24-0
C18H14N2O3S
338.381
RCKYSTKYIVULEK-UHFFFAOYSA-N
2-3-4-methanesulfinylphenyl-1-benzofuran-5-yl-5-methyl-1,3,4-oxadiazole,mmbo,1,3,4-oxadiazole, 2-methyl-5-3-4-methylsulfinyl phenyl-5-benzofuranyl,1,3,4-oxadiazole,2-methyl-5-3-4-methylsulfinyl phenyl-5-benzo b furanyl,2-3-4-methanesulfinyl phenyl-1-benzofuran-5-yl-5-methyl-1,3,4-oxadiazole,2-methyl-5-3-4-methylsulfinyl phenyl-1-benzofuran-5-yl-1,3,4-oxadiazole,2-methyl-5-3-4-methylsulfinyl phenyl-5-benzofuranyl-1,3,4-oxadiazole
24855958
2-methyl-5-[3-(4-methylsulfinylphenyl)-1-benzofuran-5-yl]-1,3,4-oxadiazole
CC1=NN=C(O1)C2=CC3=C(C=C2)OC=C3C4=CC=C(C=C4)S(=O)C

Potent GSK-3β inhibitor

Potent inhibitor of GSK-3β (IC50 = 35 nM). Oral adminstration inhibits cold water stress-induced tau hyperphosphorylation in the mouse brain.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 338.38
Percent Purity >99%
Chemical Name or Material TCS 2002

Recommended Storage : Store at +4°C