Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCS 46b, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Orally active, subtype-selective GluN1A/GluN2B (formally NR1A/NR2B) NMDA receptor antagonist (IC50 values are 5.3, 35000 and > 100000 nM for GluN1A/2B (NR1A/2B), GluN1A/2B (NR1A/2B) and GluN1A/2C (NR1A/2C) receptor subtypes respectively). Potentiates the effect of L-DOPA in 6-OHDA-lesioned rats following oral administration.
Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.
Specifications
Quantity | 50 mg |
Formula Weight | 345.44 |
Percent Purity | >98% |
Chemical Name or Material | TCS 46b |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction