Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCS HDAC6 20b, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Description
Selective inhibitor of histone deacetylase 6 (HDAC6). Inhibits HCT116 growth in combination with taxol (Cat. No. 1097). Also inhibits growth of MCF-7 cells stimulated by estrogen.
Specifications
Quantity | 10 mg |
Formula Weight | 480.7 |
Percent Purity | >98% |
Chemical Name or Material | TCS HDAC6 20b |
Safety and Handling
Recommended Storage | Store at -20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction