Tocris™ TCS PIM-1 4a

Selective, ATP-competitive Pim kinase inhibitor

Manufacturer: Tocris™ 3714/50

 View more versions of this product

Catalog No. 37-145-0R

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors TCS PIM-1 4a
    Purity >98
    Molecular Weight 273.23
    Quantity 50mg
    Product Type TCS PIM-1 4a

    Selective, ATP-competitive Pim kinase inhibitor (IC50 values are 24 and 100nM for Pim-1 and Pim-2 respectively). Displays selectivity over a panel of ~50 other kinases tested. Exhibits cytotoxicity in PC3 prostate carcinoma cells in vitro (IC50 = 17µM).

    • Chemical Name: (5Z)-5-[[3-(Trifluoromethyl)phenyl]methylene]-2,4-thiazolidinedione
    • Formula: C11H6F3NO2S
    • CAS Number: 438190-29-5
    • Solubility: Soluble to 100mM in DMSO and to 100mM in ethanol
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit