Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TDG Mouse anti-Human, DyLight 350, Clone: 1511, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243718UV

Catalog No. NBP243718UV


Add to cart

Description

Description

TDG Monoclonal antibody specifically detects TDG in Human samples. It is validated for Western Blot.
Specifications

Specifications

TDG
1511
Western Blot
Purified
TDG
Recombinant protein encompassing a sequence within the center region of human TDG. The exact sequence is proprietary.
Protein A purified
Base Excision Repair, DNA Repair
Primary
6996
Western Blot
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
EC 3.2.2.29, thymine-DNA glycosylaseG/T mismatch-specific thymine DNA glycosylase
Mouse
IgG2b
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit