Learn More
TDO2 Mouse anti-Human, Clone: OTI4G2, liquid, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner TA504730
Catalog No. 50-167-5352
Add to cart
Description
Description
Tryptophan 2, 3-dioxygenase (EC 1.13.11.11) plays a role in catalyzing the first and rat-limiting step in the kynurenine pathway, the major pathway of tryptophan metabolism.
Specifications
Specifications
TDO2
Monoclonal
Unconjugated
TDO2
Liquid
TDO2
Full length human recombit protein of human TDO2 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI4G2
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P48775
HYPTRP, TDO, TO, TPH2, TRPO
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6999