Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TDP-43/TARDBP Antibody (DB9), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250331V
Description
TDP-43/TARDBP Monoclonal specifically detects TDP-43/TARDBP in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
TDP-43/TARDBP | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TARDBP | |
A His-tagged recombinant protein corresponding to amino acid residues 208-414 of human TDP-43/TARDBP. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
DB9 | |
Western Blot, ELISA | |
ALS10TAR DNA-binding protein-43, TAR DNA binding protein, TDP43, TDP-43, TDP-43TAR DNA-binding protein 43 | |
Mouse | |
Protein A purified | |
RUO | |
23435 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction