Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TDP-43/TARDBP Mouse anti-Human, Mouse, Rat, Porcine, Bovine, Equine, Alexa Fluor 647, Clone: 3H8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192695AF647

Catalog No. NBP192695AH


Add to cart

Description

Description

TDP-43/TARDBP Monoclonal antibody specifically detects TDP-43/TARDBP in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications

Specifications

TDP-43/TARDBP
3H8
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Affinity Purified
TARDBP
Full length recombinant human TDP43 expressed in and purified from E. coli
Immunogen affinity purified
RUO
Store at 4C in the dark.
Monoclonal
Bovine, Equine, Human, Mouse, Porcine, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
ALS10TAR DNA-binding protein-43, TAR DNA binding protein, TDP43, TDP-43, TDP-43TAR DNA-binding protein 43
Mouse
IgG1
0.1 ml
Cell Cycle and Replication, Chromatin Research, DNA replication Transcription Translation and Splicing, Epigenetics, Immunology, microRNA, Mitochondrial Fusion Proteins, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Transcription Factors and Regulators, Virology Bacteria and Parasites
Primary
23435
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit