Learn More
TDP1 293T Cell Overexpression Lysate (Denatured), Abnova
Manufacturer:  Abnova Corporation H00055775T01
Catalog No. 89-002-456
Add to cart
Description
Description
Western Blotting
Specifications
Specifications
Western Blot
293T
NM_001008744
6.8
Transient overexpression cell Lysate was tested with Anti-TDP1 antibody (H00055775-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human
TDP1 293T Cell Transient Overexpression Lysate(Denatured)
66.99
pCMV-TDP1 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.