Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EAP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA550657
Description
The antibody detects endogenous levels of total TDP2 protein.
EAP2 Polyclonal antibody specifically detects EAP2 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
EAP2 | |
Polyclonal | |
Unconjugated | |
TDP2 | |
5'-Tyr-DNA phosphodiesterase; 5'-tyrosyl-DNA phosphodiesterase; AD022; AD-022; D13Ertd656e; dJ30M3.3; EAP2; EAPII; ETS1-associated protein 2; ETS1-associated protein II; hTDP2; MGC111021; MGC9099; RP1-30M3.3; Tdp2; TRAF and TNF receptor associated protein; TRAF and TNF receptor-associated protein; TTRAP; Tyr-DNA phosphodiesterase 2; tyrosyl-DNA phosphodiesterase 2; Tyrosyl-RNA phosphodiesterase; VPg unlinkase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51567, 56196 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2.6 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O95551, Q9JJX7 | |
TDP2 | |
Synthetic peptide corresponding to a region derived from internal residues of human tyrosyl-DNA phosphodiesterase 2 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction