Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TDRD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA540132
Description
Recommended positive controls: human testis tissue. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TDRD1 Polyclonal antibody specifically detects TDRD1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
TDRD1 | |
Polyclonal | |
Unconjugated | |
Tdrd1 | |
Cancer/testis antigen 41.1; CT41.1; FLJ21082; MTR-1; Tdrd1; testicular tissue protein Li 193; tudor domain containing 1; tudor domain containing protein 1; tudor domain-containing protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56165 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.32 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q9BXT4 | |
Tdrd1 | |
Recombinant protein encompassing a sequence within the center region of human TDRD1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction